Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46756515-46756778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKRP
Entrez Gene ID
79147
Gene Name
fukutin related protein
Gene Aliases
LGMD2I, MDC1C, MDDGA5, MDDGB5, MDDGC5
UniGene
Hs.515493
Gene Chromosome Location
Chr.19: 46746015-46758575 on Build GRCh38
Transcripts
NM_001039885.2 NM_024301.4 XM_005259247.2 XM_005259248.2 XM_005259249.4 XM_011527306.1 XM_011527307.1 XM_017027297.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3371390 c.1260C>G p.F420L Chr.19: 46756710-46756710 on Build GRCh38
4885259 c.1297C>A p.L433M Chr.19: 46756747-46756747 on Build GRCh38
3536208 c.1154C>T p.S385L Chr.19: 46756604-46756604 on Build GRCh38
4886188 c.1169G>T p.R390L Chr.19: 46756619-46756619 on Build GRCh38
3536209 c.1194C>T p.V398V Chr.19: 46756644-46756644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748087383 Chr.19: 46756617-46756618 on Build GRCh38
rs772020161 Chr.19: 46756635-46756636 on Build GRCh38
rs764311774 Chr.19: 46756757-46756757 on Build GRCh38
rs767570224 Chr.19: 46756640-46756640 on Build GRCh38
rs778935490 Chr.19: 46756586-46756586 on Build GRCh38
rs749787381 Chr.19: 46756536-46756536 on Build GRCh38
rs370575962 Chr.19: 46756569-46756569 on Build GRCh38
rs552260353 Chr.19: 46756590-46756590 on Build GRCh38
rs60831455 Chr.19: 46756594-46756594 on Build GRCh38
rs759140693 Chr.19: 46756562-46756562 on Build GRCh38
rs886042793 Chr.19: 46756732-46756732 on Build GRCh38
rs749652080 Chr.19: 46756607-46756607 on Build GRCh38
rs145894568 Chr.19: 46756629-46756629 on Build GRCh38
rs745314956 Chr.19: 46756612-46756612 on Build GRCh38
rs138350337 Chr.19: 46756713-46756713 on Build GRCh38
rs886043075 Chr.19: 46756537-46756537 on Build GRCh38
rs764247100 Chr.19: 46756658-46756658 on Build GRCh38
rs771708277 Chr.19: 46756617-46756617 on Build GRCh38
rs768236258 Chr.19: 46756626-46756626 on Build GRCh38
rs140217866 Chr.19: 46756586-46756586 on Build GRCh38
rs886044134 Chr.19: 46756591-46756591 on Build GRCh38
rs746533953 Chr.19: 46756703-46756703 on Build GRCh38
rs763940706 Chr.19: 46756573-46756573 on Build GRCh38
rs886043959 Chr.19: 46756706-46756707 on Build GRCh38
rs779587871 Chr.19: 46756694-46756694 on Build GRCh38
rs773049327 Chr.19: 46756724-46756724 on Build GRCh38
rs752767486 Chr.19: 46756641-46756641 on Build GRCh38
rs771388442 Chr.19: 46756547-46756547 on Build GRCh38
rs762944990 Chr.19: 46756633-46756633 on Build GRCh38
rs104894680 Chr.19: 46756604-46756604 on Build GRCh38
rs534932677 Chr.19: 46756558-46756558 on Build GRCh38
rs771793862 Chr.19: 46756565-46756565 on Build GRCh38
rs775201067 Chr.19: 46756621-46756621 on Build GRCh38
rs779577195 Chr.19: 46756610-46756610 on Build GRCh38
rs765187002 Chr.19: 46756588-46756588 on Build GRCh38
rs779487099 Chr.19: 46756674-46756674 on Build GRCh38
rs754403441 Chr.19: 46756585-46756586 on Build GRCh38
rs756347653 Chr.19: 46756595-46756595 on Build GRCh38
rs762854536 Chr.19: 46756735-46756735 on Build GRCh38
rs144236975 Chr.19: 46756756-46756756 on Build GRCh38
rs374056230 Chr.19: 46756716-46756716 on Build GRCh38
rs1031408738 Chr.19: 46756591-46756591 on Build GRCh38
rs886042367 Chr.19: 46756718-46756718 on Build GRCh38
rs747867481 Chr.19: 46756719-46756719 on Build GRCh38
rs201076863 Chr.19: 46756686-46756686 on Build GRCh38
rs376739436 Chr.19: 46756673-46756673 on Build GRCh38
rs140679502 Chr.19: 46756627-46756627 on Build GRCh38
rs376989573 Chr.19: 46756721-46756721 on Build GRCh38
rs886044083 Chr.19: 46756717-46756717 on Build GRCh38
rs756326657 Chr.19: 46756644-46756644 on Build GRCh38
rs752959830 Chr.19: 46756592-46756592 on Build GRCh38
rs769568971 Chr.19: 46756720-46756720 on Build GRCh38
rs369599106 Chr.19: 46756692-46756692 on Build GRCh38
rs373719817 Chr.19: 46756608-46756608 on Build GRCh38
rs761782258 Chr.19: 46756587-46756587 on Build GRCh38
rs751586523 Chr.19: 46756589-46756589 on Build GRCh38
rs754174318 Chr.19: 46756662-46756662 on Build GRCh38
rs772950604 Chr.19: 46756719-46756720 on Build GRCh38
rs766343049 Chr.19: 46756748-46756748 on Build GRCh38
rs369432328 Chr.19: 46756605-46756605 on Build GRCh38
rs28937904 Chr.19: 46756663-46756663 on Build GRCh38

View Full Product Details