Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 9725661-9725932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR143
Entrez Gene ID
4935
Gene Name
G protein-coupled receptor 143
Gene Aliases
NYS6, OA1
UniGene
Hs.74124
Gene Chromosome Location
Chr.X: 9725413-9765965 on Build GRCh38
Transcripts
NM_000273.2 XM_005274541.3

SNP Detail

SNP ID Chromosome Location
rs765357725 Chr.X: 9725841-9725841 on Build GRCh38
rs770745735 Chr.X: 9725804-9725804 on Build GRCh38
rs959559658 Chr.X: 9725880-9725880 on Build GRCh38
rs769263747 Chr.X: 9725783-9725783 on Build GRCh38
rs192912901 Chr.X: 9725869-9725869 on Build GRCh38
rs749808145 Chr.X: 9725825-9725825 on Build GRCh38
rs780522481 Chr.X: 9725758-9725758 on Build GRCh38
rs377013842 Chr.X: 9725806-9725806 on Build GRCh38
rs752544660 Chr.X: 9725826-9725826 on Build GRCh38
rs992308192 Chr.X: 9725910-9725910 on Build GRCh38
rs758851581 Chr.X: 9725736-9725736 on Build GRCh38
rs370069153 Chr.X: 9725822-9725822 on Build GRCh38
rs370743553 Chr.X: 9725735-9725735 on Build GRCh38
rs781539839 Chr.X: 9725775-9725775 on Build GRCh38
rs757278922 Chr.X: 9725702-9725702 on Build GRCh38
rs142625084 Chr.X: 9725805-9725805 on Build GRCh38
rs780581771 Chr.X: 9725884-9725884 on Build GRCh38
rs148263276 Chr.X: 9725768-9725768 on Build GRCh38
rs375135554 Chr.X: 9725757-9725757 on Build GRCh38
rs758630544 Chr.X: 9725871-9725871 on Build GRCh38
rs756933345 Chr.X: 9725867-9725872 on Build GRCh38
rs762544799 Chr.X: 9725798-9725798 on Build GRCh38
rs766142830 Chr.X: 9725779-9725779 on Build GRCh38
rs59194172 Chr.X: 9725879-9725879 on Build GRCh38
rs1001109766 Chr.X: 9725814-9725814 on Build GRCh38
rs145923021 Chr.X: 9725685-9725685 on Build GRCh38
rs756584912 Chr.X: 9725759-9725759 on Build GRCh38
rs1041222810 Chr.X: 9725698-9725698 on Build GRCh38
rs751266431 Chr.X: 9725728-9725729 on Build GRCh38
rs772902759 Chr.X: 9725786-9725786 on Build GRCh38
rs779314543 Chr.X: 9725728-9725728 on Build GRCh38
rs767161702 Chr.X: 9725891-9725892 on Build GRCh38
rs150992924 Chr.X: 9725828-9725828 on Build GRCh38
rs377633602 Chr.X: 9725696-9725696 on Build GRCh38

View Full Product Details