Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 6061967-6062240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL2RA
Entrez Gene ID
3559
Gene Name
interleukin 2 receptor subunit alpha
Gene Aliases
CD25, IDDM10, IL2R, IMD41, TCGFR, p55
UniGene
Hs.231367
Gene Chromosome Location
Chr.10: 6010694-6062370 on Build GRCh38
Transcripts
NM_000417.2 NM_001308242.1 NM_001308243.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6129756 c.8C>G p.S3* Chr.10: 6062144-6062144 on Build GRCh38
4958668 c.30G>A p.L10L Chr.10: 6062122-6062122 on Build GRCh38
6066251 c.39C>T p.F13F Chr.10: 6062113-6062113 on Build GRCh38
5738816 c.43A>G p.M15V Chr.10: 6062109-6062109 on Build GRCh38
1254782 c.54C>A p.G18G Chr.10: 6062098-6062098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763417993 Chr.10: 6062202-6062202 on Build GRCh38
rs750706858 Chr.10: 6062120-6062120 on Build GRCh38
rs554029632 Chr.10: 6062220-6062220 on Build GRCh38
rs537468520 Chr.10: 6062041-6062041 on Build GRCh38
rs759800570 Chr.10: 6062057-6062057 on Build GRCh38
rs75830636 Chr.10: 6062197-6062197 on Build GRCh38
rs1054097569 Chr.10: 6062074-6062074 on Build GRCh38
rs753141273 Chr.10: 6062066-6062066 on Build GRCh38
rs886041032 Chr.10: 6062088-6062091 on Build GRCh38
rs74162092 Chr.10: 6062107-6062107 on Build GRCh38
rs765489294 Chr.10: 6062040-6062040 on Build GRCh38
rs775868491 Chr.10: 6062201-6062201 on Build GRCh38
rs755829547 Chr.10: 6062151-6062151 on Build GRCh38
rs762002226 Chr.10: 6061997-6061997 on Build GRCh38
rs995688073 Chr.10: 6062209-6062209 on Build GRCh38
rs906899147 Chr.10: 6062140-6062140 on Build GRCh38
rs760443523 Chr.10: 6062191-6062191 on Build GRCh38
rs1016265267 Chr.10: 6062177-6062177 on Build GRCh38
rs539210850 Chr.10: 6062219-6062219 on Build GRCh38
rs781222262 Chr.10: 6062112-6062112 on Build GRCh38
rs779962326 Chr.10: 6062157-6062157 on Build GRCh38
rs751620727 Chr.10: 6062106-6062106 on Build GRCh38
rs747610624 Chr.10: 6062181-6062181 on Build GRCh38
rs771675867 Chr.10: 6062042-6062042 on Build GRCh38
rs267602536 Chr.10: 6062088-6062088 on Build GRCh38
rs763613912 Chr.10: 6062080-6062080 on Build GRCh38
rs992479892 Chr.10: 6062178-6062178 on Build GRCh38
rs1012971945 Chr.10: 6062117-6062117 on Build GRCh38
rs771622922 Chr.10: 6062183-6062183 on Build GRCh38
rs768519633 Chr.10: 6062172-6062172 on Build GRCh38
rs778855179 Chr.10: 6062174-6062174 on Build GRCh38
rs144690178 Chr.10: 6062170-6062170 on Build GRCh38
rs772942858 Chr.10: 6062056-6062056 on Build GRCh38
rs772713585 Chr.10: 6062188-6062188 on Build GRCh38
rs916932655 Chr.10: 6062216-6062216 on Build GRCh38
rs1042060738 Chr.10: 6061996-6061996 on Build GRCh38
rs764712311 Chr.10: 6062096-6062096 on Build GRCh38

View Full Product Details