The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016350.4 | NP_057434.4 | 26 - 27 | 3821 | 145 |
NM_020921.3 | NP_065972.3 | 27 - 28 | 5960 | 145 | |
NM_182944.2 | NP_891989.2 | 27 - 28 | 5960 | 145 | |
NM_182946.1 | NP_891991.1 | 27 - 28 | 5960 | 145 | |
XM_011536817.1 | XP_011535119.1 | 27 - 28 | 5934 | 145 | |
XM_011536819.2 | XP_011535121.1 | 27 - 28 | 5934 | 145 | |
XM_011536822.1 | XP_011535124.1 | 26 - 27 | 3795 | 145 | |
XM_011536823.2 | XP_011535125.1 | 26 - 27 | 5811 | 145 | |
GenBank mRNA | AB046785.2 | - | 26 - 27 | 3743 | 145 |
AF186776.1 | - | 8 - 9 | 1089 | 145 | |
AF212162.2 | - | 26 - 27 | 5989 | 145 | |
AF223937.1 | - | 28 - 29 | 5919 | 145 | |
AF223938.1 | - | 28 - 29 | 5919 | 145 | |
AF223939.1 | - | 28 - 29 | 5919 | 145 | |
AF302773.2 | - | 27 - 28 | 5960 | 145 | |
AK025652.1 | - | 8 - 9 | 1080 | 145 | |
AK026083.1 | - | 10 - 11 | 1519 | 145 | |
AY027794.1 | - | 8 - 9 | 1089 | 145 | |
AY027795.1 | - | 8 - 9 | 1089 | 145 | |
AY027796.1 | - | 8 - 9 | 1089 | 145 | |
AY736182.1 | - | 27 - 28 | 5960 | 145 | |
AY736183.1 | - | 2 - 3 | 222 | 145 | |
BC034708.2 | - | 1 - 2 | 142 | 145 | |
BC090932.1 | - | 24 - 25 | 3665 | 145 |