Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 86578668-86578899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGB3
Entrez Gene ID
54714
Gene Name
cyclic nucleotide gated channel beta 3
Gene Aliases
ACHM1
UniGene
Hs.154433
Gene Chromosome Location
Chr.8: 86573608-86743675 on Build GRCh38
Transcripts
NM_019098.4 XM_011517138.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
317127 c.1948G>T p.A650S Chr.8: 86578844-86578844 on Build GRCh38
7100492 c.1944G>A p.Q648Q Chr.8: 86578848-86578848 on Build GRCh38
3367417 c.2036T>C p.L679P Chr.8: 86578756-86578756 on Build GRCh38
6549506 c.1969C>A p.P657T Chr.8: 86578823-86578823 on Build GRCh38
2789791 c.1957G>A p.A653T Chr.8: 86578835-86578835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780866922 Chr.8: 86578807-86578808 on Build GRCh38
rs143875055 Chr.8: 86578862-86578862 on Build GRCh38
rs774779115 Chr.8: 86578828-86578828 on Build GRCh38
rs745557293 Chr.8: 86578855-86578855 on Build GRCh38
rs375843177 Chr.8: 86578775-86578775 on Build GRCh38
rs926474552 Chr.8: 86578783-86578783 on Build GRCh38
rs756335664 Chr.8: 86578756-86578756 on Build GRCh38
rs142347723 Chr.8: 86578788-86578788 on Build GRCh38
rs767430685 Chr.8: 86578836-86578836 on Build GRCh38
rs766831491 Chr.8: 86578732-86578732 on Build GRCh38
rs535555046 Chr.8: 86578765-86578765 on Build GRCh38
rs760845597 Chr.8: 86578851-86578851 on Build GRCh38
rs750257554 Chr.8: 86578864-86578864 on Build GRCh38
rs771447950 Chr.8: 86578824-86578824 on Build GRCh38
rs746605655 Chr.8: 86578793-86578793 on Build GRCh38
rs751631816 Chr.8: 86578734-86578734 on Build GRCh38
rs192448853 Chr.8: 86578706-86578706 on Build GRCh38
rs370862624 Chr.8: 86578839-86578839 on Build GRCh38
rs759584325 Chr.8: 86578741-86578741 on Build GRCh38
rs144474033 Chr.8: 86578745-86578745 on Build GRCh38
rs768447937 Chr.8: 86578798-86578798 on Build GRCh38
rs1057517388 Chr.8: 86578865-86578865 on Build GRCh38
rs948664388 Chr.8: 86578866-86578866 on Build GRCh38
rs753607658 Chr.8: 86578860-86578860 on Build GRCh38
rs753663511 Chr.8: 86578767-86578767 on Build GRCh38
rs749714532 Chr.8: 86578815-86578815 on Build GRCh38
rs758366612 Chr.8: 86578867-86578867 on Build GRCh38
rs868399385 Chr.8: 86578743-86578744 on Build GRCh38
rs147991883 Chr.8: 86578785-86578785 on Build GRCh38
rs761969118 Chr.8: 86578781-86578781 on Build GRCh38
rs773675958 Chr.8: 86578802-86578802 on Build GRCh38
rs377730576 Chr.8: 86578705-86578705 on Build GRCh38
rs752985345 Chr.8: 86578751-86578751 on Build GRCh38
rs149119556 Chr.8: 86578856-86578856 on Build GRCh38
rs368249894 Chr.8: 86578835-86578835 on Build GRCh38
rs971820132 Chr.8: 86578823-86578823 on Build GRCh38
rs954285823 Chr.8: 86578746-86578746 on Build GRCh38
rs146161333 Chr.8: 86578789-86578789 on Build GRCh38
rs200892871 Chr.8: 86578753-86578753 on Build GRCh38

View Full Product Details