Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM189A2
Entrez Gene ID
9413
Gene Name
family with sequence similarity 189 member A2
Gene Aliases
C9orf61, X123
Chromosome Location
Chr.9: 69324572 - 69392461 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127608.1 NP_001121080.1 10 - 11 1246 64
NM_004816.3 NP_004807.3 10 - 11 1562 64
XM_005252307.4 XP_005252364.1 10 - 11 2123 64
XM_017015324.1 XP_016870813.1 9 - 10 1307 64
GenBank mRNA AK056152.1 - 8 - 9 1190 64
AK126127.1 - 10 - 11 1562 64
BC113683.1 - 10 - 11 1205 64
BC113685.1 - 10 - 11 1205 64
BX641153.1 - 6 - 7 8627 64
L27479.1 - 1 - 2 529 64

View Full Product Details