The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278623.1 | NP_001265552.1 | 7 - 8 | 1048 | 116 |
NM_001278624.1 | NP_001265553.1 | 7 - 8 | 1186 | 116 | |
NM_152995.5 | NP_694540.3 | 7 - 8 | 1088 | 116 | |
NR_103795.1 | 7 - 8 | 1048 | 116 | ||
GenBank mRNA | AB085695.1 | - | 7 - 8 | 1044 | 116 |
AB181916.1 | - | 7 - 8 | 1168 | 116 | |
AK131285.1 | - | 5 - 6 | 634 | 116 | |
AK302440.1 | - | 7 - 8 | 1046 | 116 | |
BC051193.1 | - | 7 - 8 | 1019 | 116 |