Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77210557-77210782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS13A
Entrez Gene ID
23230
Gene Name
vacuolar protein sorting 13 homolog A
Gene Aliases
CHAC, CHOREIN
UniGene
Hs.459790
Gene Chromosome Location
Chr.9: 77177353-77421537 on Build GRCh38
Transcripts
NM_001018037.1 NM_001018038.2 NM_015186.3 NM_033305.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3217985 c.506G>A p.R169Q Chr.9: 77210626-77210626 on Build GRCh38
7287278 c.496-2A>C p.? Chr.9: 77210614-77210614 on Build GRCh38
3217984 c.506G>A p.R169Q Chr.9: 77210626-77210626 on Build GRCh38
6903139 c.507G>A p.R169R Chr.9: 77210627-77210627 on Build GRCh38
6903141 c.507G>A p.R169R Chr.9: 77210627-77210627 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781084069 Chr.9: 77210726-77210726 on Build GRCh38
rs879199752 Chr.9: 77210695-77210695 on Build GRCh38
rs768562676 Chr.9: 77210613-77210613 on Build GRCh38
rs779057916 Chr.9: 77210704-77210704 on Build GRCh38
rs371449823 Chr.9: 77210717-77210717 on Build GRCh38
rs760037642 Chr.9: 77210654-77210654 on Build GRCh38
rs766587340 Chr.9: 77210639-77210639 on Build GRCh38
rs753300394 Chr.9: 77210679-77210679 on Build GRCh38
rs141109675 Chr.9: 77210636-77210636 on Build GRCh38
rs139380985 Chr.9: 77210625-77210625 on Build GRCh38
rs1027682562 Chr.9: 77210735-77210735 on Build GRCh38
rs952979414 Chr.9: 77210741-77210741 on Build GRCh38
rs772689261 Chr.9: 77210635-77210635 on Build GRCh38
rs544104609 Chr.9: 77210608-77210608 on Build GRCh38
rs972083958 Chr.9: 77210653-77210653 on Build GRCh38
rs370541609 Chr.9: 77210607-77210607 on Build GRCh38
rs768135367 Chr.9: 77210666-77210666 on Build GRCh38
rs774869789 Chr.9: 77210651-77210651 on Build GRCh38
rs768517006 Chr.9: 77210694-77210694 on Build GRCh38
rs369548882 Chr.9: 77210727-77210727 on Build GRCh38
rs757482674 Chr.9: 77210709-77210709 on Build GRCh38
rs764115216 Chr.9: 77210599-77210602 on Build GRCh38
rs546023010 Chr.9: 77210716-77210716 on Build GRCh38
rs146334644 Chr.9: 77210696-77210696 on Build GRCh38
rs756128729 Chr.9: 77210687-77210687 on Build GRCh38
rs769393043 Chr.9: 77210626-77210626 on Build GRCh38
rs367807671 Chr.9: 77210693-77210693 on Build GRCh38
rs764260920 Chr.9: 77210689-77210689 on Build GRCh38
rs756970615 Chr.9: 77210715-77210715 on Build GRCh38
rs751582738 Chr.9: 77210709-77210710 on Build GRCh38

View Full Product Details