Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46672404-46672629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS35
Entrez Gene ID
55737
Gene Name
VPS35 retromer complex component
Gene Aliases
MEM3, PARK17
UniGene
Hs.454528
Gene Chromosome Location
Chr.16: 46657979-46689232 on Build GRCh38
Transcripts
NM_018206.5 XM_005256045.3 XM_011523227.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5770714 c.1184C>A p.S395* Chr.16: 46672449-46672449 on Build GRCh38
390509 c.1161-2A>T p.? Chr.16: 46672474-46672474 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs906414936 Chr.16: 46672595-46672595 on Build GRCh38
rs754137147 Chr.16: 46672453-46672453 on Build GRCh38
rs779224141 Chr.16: 46672456-46672457 on Build GRCh38
rs757485744 Chr.16: 46672463-46672463 on Build GRCh38
rs373540036 Chr.16: 46672520-46672520 on Build GRCh38
rs183971771 Chr.16: 46672542-46672542 on Build GRCh38
rs779065298 Chr.16: 46672495-46672495 on Build GRCh38
rs191349966 Chr.16: 46672559-46672559 on Build GRCh38
rs764365507 Chr.16: 46672438-46672438 on Build GRCh38
rs745425051 Chr.16: 46672518-46672518 on Build GRCh38
rs750539567 Chr.16: 46672504-46672504 on Build GRCh38
rs1042064582 Chr.16: 46672581-46672581 on Build GRCh38
rs560831455 Chr.16: 46672543-46672543 on Build GRCh38
rs746383321 Chr.16: 46672477-46672477 on Build GRCh38

View Full Product Details