Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41257434-41257683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UCHL1
Entrez Gene ID
7345
Gene Name
ubiquitin C-terminal hydrolase L1
Gene Aliases
HEL-117, HEL-S-53, NDGOA, PARK5, PGP 9.5, PGP9.5, PGP95, Uch-L1
UniGene
Hs.518731
Gene Chromosome Location
Chr.4: 41256881-41268429 on Build GRCh38
Transcripts
NM_004181.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4159070 c.53C>A p.S18Y Chr.4: 41257616-41257616 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778151038 Chr.4: 41257596-41257596 on Build GRCh38
rs758721522 Chr.4: 41257593-41257593 on Build GRCh38
rs767261015 Chr.4: 41257623-41257623 on Build GRCh38
rs556171924 Chr.4: 41257468-41257468 on Build GRCh38
rs747510594 Chr.4: 41257603-41257603 on Build GRCh38
rs769924890 Chr.4: 41257618-41257618 on Build GRCh38
rs919210760 Chr.4: 41257559-41257559 on Build GRCh38
rs773342302 Chr.4: 41257572-41257573 on Build GRCh38
rs985260501 Chr.4: 41257524-41257524 on Build GRCh38
rs565148227 Chr.4: 41257499-41257499 on Build GRCh38
rs541220563 Chr.4: 41257550-41257550 on Build GRCh38
rs1031726237 Chr.4: 41257459-41257459 on Build GRCh38
rs771560835 Chr.4: 41257605-41257605 on Build GRCh38
rs544988955 Chr.4: 41257490-41257490 on Build GRCh38
rs930709004 Chr.4: 41257547-41257547 on Build GRCh38
rs769879042 Chr.4: 41257573-41257573 on Build GRCh38
rs1049115291 Chr.4: 41257576-41257576 on Build GRCh38
rs766292262 Chr.4: 41257630-41257630 on Build GRCh38
rs955752700 Chr.4: 41257460-41257460 on Build GRCh38
rs559788950 Chr.4: 41257544-41257544 on Build GRCh38
rs868560463 Chr.4: 41257654-41257654 on Build GRCh38
rs5030732 Chr.4: 41257616-41257616 on Build GRCh38
rs764766596 Chr.4: 41257662-41257662 on Build GRCh38
rs972692048 Chr.4: 41257552-41257552 on Build GRCh38
rs909743752 Chr.4: 41257538-41257538 on Build GRCh38
rs894583471 Chr.4: 41257594-41257594 on Build GRCh38
rs760676226 Chr.4: 41257628-41257628 on Build GRCh38
rs754490470 Chr.4: 41257657-41257657 on Build GRCh38
rs999158009 Chr.4: 41257648-41257648 on Build GRCh38
rs753345185 Chr.4: 41257566-41257566 on Build GRCh38
rs747771091 Chr.4: 41257573-41257573 on Build GRCh38
rs1045884870 Chr.4: 41257626-41257626 on Build GRCh38
rs745957339 Chr.4: 41257617-41257617 on Build GRCh38
rs763182177 Chr.4: 41257619-41257619 on Build GRCh38
rs112409507 Chr.4: 41257481-41257481 on Build GRCh38
rs753720793 Chr.4: 41257632-41257632 on Build GRCh38
rs372937139 Chr.4: 41257569-41257569 on Build GRCh38
rs1029422646 Chr.4: 41257494-41257494 on Build GRCh38
rs368554502 Chr.4: 41257577-41257577 on Build GRCh38
rs754687056 Chr.4: 41257568-41257568 on Build GRCh38
rs953833361 Chr.4: 41257506-41257506 on Build GRCh38
rs560256408 Chr.4: 41257555-41257555 on Build GRCh38
rs929329652 Chr.4: 41257560-41257560 on Build GRCh38
rs370928430 Chr.4: 41257620-41257620 on Build GRCh38

View Full Product Details