The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276282.3 | NP_001263211.1 | 4 - 5 | 1002 | 112 |
NM_207303.4 | NP_997186.1 | 4 - 5 | 1002 | 112 | |
XM_011539587.1 | XP_011537889.1 | 4 - 5 | 997 | 112 | |
XM_011539588.2 | XP_011537890.1 | 4 - 5 | 547 | 112 | |
XM_011539589.2 | XP_011537891.1 | 4 - 5 | 997 | 112 | |
XM_011539590.2 | XP_011537892.1 | 4 - 5 | 2899 | 112 | |
XM_011539591.2 | XP_011537893.1 | 4 - 5 | 870 | 112 | |
XM_017016035.1 | XP_016871524.1 | 4 - 5 | 997 | 112 | |
XM_017016036.1 | XP_016871525.1 | 4 - 5 | 997 | 112 | |
XM_017016037.1 | XP_016871526.1 | 3 - 4 | 257 | 112 | |
XM_017016038.1 | XP_016871527.1 | 4 - 5 | 8209 | 112 | |
GenBank mRNA | AK127277.1 | - | 4 - 5 | 480 | 112 |
AY442317.1 | - | 4 - 5 | 616 | 112 | |
BC035157.1 | - | 4 - 5 | 1008 | 112 | |
BC047716.1 | - | 4 - 5 | 947 | 112 | |
BC139916.1 | - | 4 - 5 | 991 | 112 | |
BC139923.1 | - | 4 - 5 | 991 | 112 |