Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATRNL1
Entrez Gene ID
26033
Gene Name
attractin like 1
Gene Aliases
ALP, bA338L11.1, bA454H24.1
Chromosome Location
Chr.10: 115093365 - 115948999 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276282.3 NP_001263211.1 7 - 8 1473 97
NM_207303.4 NP_997186.1 7 - 8 1473 97
NR_074088.2 9 - 10 1363 97
XM_011539587.1 XP_011537889.1 7 - 8 1468 97
XM_011539588.2 XP_011537890.1 7 - 8 1018 97
XM_011539589.2 XP_011537891.1 7 - 8 1468 97
XM_011539590.2 XP_011537892.1 7 - 8 3370 97
XM_011539591.2 XP_011537893.1 7 - 8 1341 97
XM_017016035.1 XP_016871524.1 7 - 8 1468 97
XM_017016036.1 XP_016871525.1 7 - 8 1468 97
GenBank mRNA AB011106.2 - 1 - 2 34 97
AK127277.1 - 7 - 8 951 97
AY442317.1 - 7 - 8 1087 97
BC035157.1 - 7 - 8 1479 97
BC047716.1 - 7 - 8 1418 97
BC139916.1 - 7 - 8 1462 97
BC139923.1 - 7 - 8 1462 97

View Full Product Details