Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68211754-68211989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLN6
Entrez Gene ID
54982
Gene Name
ceroid-lipofuscinosis, neuronal 6, late infantile, variant
Gene Aliases
CLN4A, HsT18960, nclf
UniGene
Hs.584921
Gene Chromosome Location
Chr.15: 68206992-68229742 on Build GRCh38
Transcripts
NM_017882.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3678253 c.298-6C>T p.? Chr.15: 68211869-68211869 on Build GRCh38
6077863 c.317G>T p.R106L Chr.15: 68211844-68211844 on Build GRCh38
3503448 c.403C>T p.H135Y Chr.15: 68211758-68211758 on Build GRCh38
6299936 c.339G>A p.T113T Chr.15: 68211822-68211822 on Build GRCh38
5054441 c.316delC p.R106fs*10 Chr.15: 68211845-68211845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769747737 Chr.15: 68211847-68211847 on Build GRCh38
rs762232673 Chr.15: 68211818-68211818 on Build GRCh38
rs755184214 Chr.15: 68211840-68211840 on Build GRCh38
rs185601303 Chr.15: 68211920-68211920 on Build GRCh38
rs763431994 Chr.15: 68211897-68211897 on Build GRCh38
rs397515352 Chr.15: 68211844-68211845 on Build GRCh38
rs767729558 Chr.15: 68211897-68211898 on Build GRCh38
rs777921628 Chr.15: 68211850-68211850 on Build GRCh38
rs752212030 Chr.15: 68211811-68211811 on Build GRCh38
rs952586818 Chr.15: 68211880-68211880 on Build GRCh38
rs766756538 Chr.15: 68211908-68211908 on Build GRCh38
rs1053920744 Chr.15: 68211812-68211812 on Build GRCh38
rs143928329 Chr.15: 68211833-68211833 on Build GRCh38
rs1046423746 Chr.15: 68211791-68211791 on Build GRCh38
rs104894484 Chr.15: 68211793-68211793 on Build GRCh38
rs140179213 Chr.15: 68211832-68211832 on Build GRCh38
rs758830997 Chr.15: 68211823-68211823 on Build GRCh38
rs759241838 Chr.15: 68211806-68211806 on Build GRCh38
rs935340832 Chr.15: 68211935-68211936 on Build GRCh38
rs144808715 Chr.15: 68211894-68211894 on Build GRCh38
rs146135801 Chr.15: 68211822-68211822 on Build GRCh38
rs201095412 Chr.15: 68211854-68211854 on Build GRCh38
rs924518551 Chr.15: 68211916-68211916 on Build GRCh38
rs796052353 Chr.15: 68211839-68211839 on Build GRCh38
rs908457846 Chr.15: 68211918-68211918 on Build GRCh38
rs202226970 Chr.15: 68211845-68211845 on Build GRCh38
rs148547876 Chr.15: 68211798-68211798 on Build GRCh38
rs886051447 Chr.15: 68211804-68211806 on Build GRCh38
rs774238261 Chr.15: 68211860-68211860 on Build GRCh38
rs781133812 Chr.15: 68211844-68211844 on Build GRCh38
rs995775001 Chr.15: 68211828-68211828 on Build GRCh38
rs982110599 Chr.15: 68211931-68211931 on Build GRCh38
rs760292374 Chr.15: 68211886-68211886 on Build GRCh38
rs914416326 Chr.15: 68211879-68211879 on Build GRCh38
rs1007902706 Chr.15: 68211829-68211829 on Build GRCh38
rs765719755 Chr.15: 68211819-68211819 on Build GRCh38
rs368661456 Chr.15: 68211878-68211878 on Build GRCh38
rs771689244 Chr.15: 68211876-68211876 on Build GRCh38
rs796052352 Chr.15: 68211857-68211857 on Build GRCh38
rs154774634 Chr.15: 68211853-68211853 on Build GRCh38
rs955286997 Chr.15: 68211842-68211842 on Build GRCh38
rs975223501 Chr.15: 68211846-68211846 on Build GRCh38
rs772893554 Chr.15: 68211788-68211788 on Build GRCh38
rs117038427 Chr.15: 68211869-68211869 on Build GRCh38
rs970668097 Chr.15: 68211924-68211924 on Build GRCh38
rs141950483 Chr.15: 68211804-68211804 on Build GRCh38
rs751850377 Chr.15: 68211913-68211913 on Build GRCh38
rs926709197 Chr.15: 68211952-68211952 on Build GRCh38

View Full Product Details