The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281496.1 | NP_001268425.1 | 7 - 7 | 1336 | 155 |
NM_001281497.1 | NP_001268426.1 | 6 - 6 | 937 | 155 | |
NM_001281715.1 | NP_001268644.1 | 8 - 8 | 1380 | 155 | |
NM_002634.3 | NP_002625.1 | 7 - 7 | 1288 | 155 | |
XM_017024763.1 | XP_016880252.1 | 8 - 8 | 1430 | 155 | |
GenBank mRNA | AK223181.1 | - | 7 - 7 | 1263 | 155 |
BP267961.1 | - | 1 - 1 | 171 | 155 | |
BQ574485.1 | - | 1 - 1 | 155 |