The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002916.4 | NP_001002916.3 | 1 - 2 | 519 | 142 |
GenBank mRNA | BC038109.2 | - | 1 - 2 | 516 | 142 |
BC118604.1 | - | 1 - 2 | 512 | 142 | |
BC121816.1 | - | 1 - 2 | 512 | 142 |