The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000762.5 | NP_000753.3 | 9 - 9 | 1560 | 131 |
GenBank mRNA | BC096253.3 | - | 9 - 9 | 131 | |
BC096254.1 | - | 9 - 9 | 1548 | 131 | |
BC096255.1 | - | 9 - 9 | 1548 | 131 | |
BC096256.1 | - | 9 - 9 | 1548 | 131 | |
M33318.1 | - | 9 - 9 | 1548 | 131 | |
X06401.1 | - | 5 - 5 | 772 | 131 | |
X13897.1 | - | 9 - 9 | 1534 | 131 |