Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF6
Entrez Gene ID
221458
Gene Name
kinesin family member 6
Gene Aliases
C6orf102, dJ1043E3.1, dJ137F1.4, dJ188D3.1
Chromosome Location
Chr.6: 39335100 - 39725405 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289020.1 NP_001275949.1 16 - 17 1986 70
NM_001289021.1 NP_001275950.1 16 - 17 1869 70
NM_001289024.1 NP_001275953.1 4 - 5 510 70
NM_145027.4 NP_659464.3 17 - 18 2037 70
XM_005248904.4 XP_005248961.1 17 - 18 2037 70
XM_011514357.2 XP_011512659.1 17 - 18 1962 70
XM_011514358.2 XP_011512660.1 17 - 18 1962 70
XM_011514359.2 XP_011512661.1 17 - 18 1962 70
XM_017010427.1 XP_016865916.1 16 - 17 1927 70
XM_017010428.1 XP_016865917.1 12 - 13 1387 70
GenBank mRNA AK123217.1 - 4 - 5 510 70
AK131471.1 - 17 - 18 2037 70
AL832634.2 - 4 - 5 273 70
BC022074.2 - 4 - 5 451 70
BC098418.1 - 4 - 5 469 70
BC117307.1 - 16 - 17 1799 70
BC143444.1 - 16 - 17 1916 70
BC143445.1 - 16 - 17 1799 70
BX649045.1 - 14 - 15 1617 70
DA167995.1 - 4 - 5 510 70

View Full Product Details