The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173689.6 | NP_775960.4 | 11 - 12 | 3602 | 123 |
NR_104603.1 | 6 - 7 | 2622 | 123 | ||
XM_005251934.2 | XP_005251991.1 | 6 - 7 | 2622 | 123 | |
XM_011518556.2 | XP_011516858.1 | 11 - 12 | 3575 | 123 | |
XM_011518557.2 | XP_011516859.1 | 11 - 12 | 3604 | 123 | |
XM_011518558.2 | XP_011516860.1 | 12 - 13 | 3964 | 123 | |
GenBank mRNA | AK095783.1 | - | 2 - 3 | 239 | 123 |
AK126775.1 | - | 6 - 7 | 2622 | 123 | |
AY720432.1 | - | 11 - 12 | 3508 | 123 |