The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173689.6 | NP_775960.4 | 1 - 2 | 192 | 77 |
XM_011518556.2 | XP_011516858.1 | 1 - 2 | 192 | 77 | |
GenBank mRNA | AK123000.1 | - | 1 - 2 | 182 | 77 |
AY720432.1 | - | 1 - 2 | 98 | 77 | |
DA240339.1 | - | 1 - 2 | 185 | 77 |