The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000819.4 | NP_000810.1 | 2 - 3 | 282 | 71 |
NM_001136005.1 | NP_001129477.1 | 2 - 3 | 458 | 71 | |
NM_001136006.1 | NP_001129478.1 | 2 - 3 | 421 | 71 | |
NM_175085.2 | NP_780294.1 | 2 - 3 | 282 | 71 | |
XM_005260941.1 | XP_005260998.1 | 2 - 3 | 558 | 71 | |
XM_006723989.1 | XP_006724052.1 | 2 - 3 | 500 | 71 | |
XM_006723990.1 | XP_006724053.1 | 2 - 3 | 388 | 71 | |
XM_011529526.1 | XP_011527828.1 | 2 - 3 | 467 | 71 | |
GenBank mRNA | AB208785.1 | - | 2 - 3 | 348 | 71 |
AK292560.1 | - | 2 - 3 | 397 | 71 | |
AK292897.1 | - | 2 - 3 | 235 | 71 | |
BC038958.1 | - | 2 - 3 | 245 | 71 | |
BC093641.1 | - | 1 - 2 | 201 | 71 | |
BC101565.1 | - | 1 - 2 | 201 | 71 | |
BC107712.1 | - | 2 - 3 | 236 | 71 | |
BM172468.1 | - | 2 - 3 | 71 | ||
BX648124.1 | - | 2 - 3 | 399 | 71 | |
D32051.1 | - | 2 - 3 | 218 | 71 | |
DB062974.1 | - | 2 - 3 | 421 | 71 | |
DB236855.1 | - | 2 - 3 | 282 | 71 | |
X54199.1 | - | 2 - 3 | 225 | 71 |