Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC37A4
Entrez Gene ID
2542
Gene Name
solute carrier family 37 member 4
Gene Aliases
G6PT1, G6PT2, G6PT3, GSD1b, GSD1c, GSD1d, PRO0685, TRG-19, TRG19
Chromosome Location
Chr.11: 119024351 - 119030906 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164277.1 NP_001157749.1 6 - 7 1538 107
NM_001164278.1 NP_001157750.1 6 - 7 1222 107
NM_001164279.1 NP_001157751.1 6 - 7 964 107
NM_001164280.1 NP_001157752.1 4 - 5 980 107
NM_001467.5 NP_001458.1 5 - 6 1034 107
GenBank mRNA AF110819.1 - 4 - 5 781 107
AF110820.1 - 4 - 5 781 107
AF110822.1 - 4 - 5 781 107
AF111852.1 - 2 - 3 1534 107
AK289642.1 - 6 - 7 1506 107
AK298395.1 - 6 - 7 918 107
AK300646.1 - 6 - 7 930 107
AK312628.1 - 6 - 7 1204 107
AY423732.1 - 4 - 5 801 107
BC002400.2 - 4 - 5 982 107
BC003589.1 - 4 - 5 982 107
BC014663.1 - 4 - 5 989 107
BC015650.2 - 5 - 6 1454 107
BC064563.1 - 4 - 5 977 107
CR456733.1 - 4 - 5 781 107
Y15409.1 - 4 - 5 950 107

View Full Product Details