Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC37A4
Entrez Gene ID
2542
Gene Name
solute carrier family 37 member 4
Gene Aliases
G6PT1, G6PT2, G6PT3, GSD1b, GSD1c, GSD1d, PRO0685, TRG-19, TRG19
Chromosome Location
Chr.11: 119024351 - 119030906 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164279.1 NP_001157751.1 1 - 2 56 88
NM_001467.5 NP_001458.1 1 - 2 56 88
GenBank mRNA DC295438.1 - 1 - 2 56 88

View Full Product Details