The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012112.4 | NP_036244.2 | 12 - 13 | 2106 | 73 |
XM_011528697.2 | XP_011526999.1 | 12 - 13 | 1664 | 73 | |
XM_011528699.2 | XP_011527001.1 | 11 - 12 | 1852 | 73 | |
XM_011528700.2 | XP_011527002.1 | 11 - 12 | 1585 | 73 | |
GenBank mRNA | AB024704.1 | - | 12 - 13 | 1879 | 73 |
AB027467.1 | - | 11 - 12 | 1515 | 73 | |
AF098158.1 | - | 12 - 13 | 1899 | 73 | |
AF146731.1 | - | 11 - 12 | 1512 | 73 | |
AF244547.1 | - | 9 - 10 | 1286 | 73 | |
AF287265.1 | - | 13 - 14 | 1980 | 73 | |
AK001391.1 | - | 8 - 9 | 1189 | 73 | |
AK312549.1 | - | 12 - 13 | 1884 | 73 | |
AL117534.1 | - | 10 - 11 | 1350 | 73 | |
AY734680.1 | - | 10 - 11 | 1408 | 73 | |
BC004136.1 | - | 12 - 13 | 1836 | 73 | |
BC020207.1 | - | 12 - 13 | 1865 | 73 |