The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012112.4 | NP_036244.2 | 16 - 17 | 2646 | 80 |
XM_011528697.2 | XP_011526999.1 | 16 - 17 | 2204 | 80 | |
XM_011528699.2 | XP_011527001.1 | 15 - 16 | 2392 | 80 | |
GenBank mRNA | AB024704.1 | - | 16 - 17 | 2419 | 80 |
AB027467.1 | - | 15 - 16 | 2055 | 80 | |
AF098158.1 | - | 16 - 17 | 2439 | 80 | |
AF146731.1 | - | 15 - 16 | 2052 | 80 | |
AF244547.1 | - | 13 - 14 | 1826 | 80 | |
AF287265.1 | - | 17 - 18 | 2520 | 80 | |
AK001391.1 | - | 12 - 13 | 1729 | 80 | |
AK093379.1 | - | 5 - 6 | 1120 | 80 | |
AK312549.1 | - | 16 - 17 | 2424 | 80 | |
AL117534.1 | - | 14 - 15 | 1890 | 80 | |
AY734680.1 | - | 14 - 15 | 1948 | 80 | |
BC004136.1 | - | 16 - 17 | 2376 | 80 | |
BC011357.1 | - | 3 - 4 | 348 | 80 | |
BC020207.1 | - | 16 - 17 | 2405 | 80 |