The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012112.4 | NP_036244.2 | 4 - 5 | 931 | 129 |
XM_011528697.2 | XP_011526999.1 | 4 - 5 | 489 | 129 | |
XM_011528699.2 | XP_011527001.1 | 3 - 4 | 677 | 129 | |
XM_011528700.2 | XP_011527002.1 | 3 - 4 | 410 | 129 | |
GenBank mRNA | AB024704.1 | - | 4 - 5 | 704 | 129 |
AB027467.1 | - | 3 - 4 | 340 | 129 | |
AF098158.1 | - | 4 - 5 | 724 | 129 | |
AF146731.1 | - | 3 - 4 | 337 | 129 | |
AF244547.1 | - | 1 - 2 | 111 | 129 | |
AF287265.1 | - | 4 - 5 | 697 | 129 | |
AK312549.1 | - | 4 - 5 | 709 | 129 | |
AL117534.1 | - | 2 - 3 | 175 | 129 | |
AY734680.1 | - | 2 - 3 | 233 | 129 | |
BC004136.1 | - | 4 - 5 | 661 | 129 | |
BC020207.1 | - | 4 - 5 | 690 | 129 |