The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077700.2 | NP_001071168.2 | 10 - 11 | 1140 | 158 |
NM_001077701.2 | NP_001071169.1 | 9 - 10 | 996 | 158 | |
NM_001077702.2 | NP_001071170.2 | 11 - 12 | 1214 | 158 | |
NM_001077703.2 | NP_001071171.2 | 10 - 11 | 1140 | 158 | |
NM_001077704.2 | NP_001071172.1 | 9 - 10 | 996 | 158 | |
NM_001146110.1 | NP_001139582.1 | 11 - 12 | 1217 | 158 | |
NM_001146111.1 | NP_001139583.1 | 11 - 12 | 1214 | 158 | |
NM_001146112.1 | NP_001139584.1 | 11 - 12 | 1175 | 158 | |
NM_001146113.1 | NP_001139585.1 | 9 - 10 | 994 | 158 | |
NM_020948.3 | NP_065999.2 | 11 - 12 | 1214 | 158 | |
XM_005271076.3 | XP_005271133.1 | 11 - 12 | 1252 | 158 | |
XM_011541864.2 | XP_011540166.1 | 12 - 13 | 1159 | 158 | |
XM_011541865.2 | XP_011540167.1 | 12 - 13 | 1120 | 158 | |
XM_017001919.1 | XP_016857408.1 | 12 - 13 | 1136 | 158 | |
XM_017001920.1 | XP_016857409.1 | 12 - 13 | 1153 | 158 | |
XM_017001921.1 | XP_016857410.1 | 12 - 13 | 1181 | 158 | |
XM_017001922.1 | XP_016857411.1 | 12 - 13 | 1156 | 158 | |
XM_017001923.1 | XP_016857412.1 | 10 - 11 | 976 | 158 | |
XM_017001924.1 | XP_016857413.1 | 11 - 12 | 1108 | 158 | |
XM_017001925.1 | XP_016857414.1 | 11 - 12 | 1213 | 158 | |
XM_017001926.1 | XP_016857415.1 | 12 - 13 | 1152 | 158 | |
XM_017001927.1 | XP_016857416.1 | 10 - 11 | 1025 | 158 | |
XM_017001928.1 | XP_016857417.1 | 10 - 11 | 1101 | 158 | |
XM_017001929.1 | XP_016857418.1 | 11 - 12 | 1082 | 158 | |
XM_017001930.1 | XP_016857419.1 | 10 - 11 | 1026 | 158 | |
XM_017001931.1 | XP_016857420.1 | 11 - 12 | 1060 | 158 | |
XM_017001932.1 | XP_016857421.1 | 9 - 10 | 989 | 158 | |
XM_017001933.1 | XP_016857422.1 | 8 - 9 | 843 | 158 | |
XM_017001934.1 | XP_016857423.1 | 7 - 8 | 739 | 158 | |
GenBank mRNA | AB046830.1 | - | 7 - 8 | 765 | 158 |
AF515446.1 | - | 9 - 10 | 996 | 158 | |
AF515447.1 | - | 10 - 11 | 1077 | 158 | |
AF515448.2 | - | 11 - 12 | 1151 | 158 | |
AK302061.1 | - | 10 - 11 | 1140 | 158 | |
AK307717.1 | - | 11 - 12 | 1111 | 158 | |
AL831987.2 | - | 11 - 12 | 1156 | 158 | |
AY124186.2 | - | 11 - 12 | 1151 | 158 | |
AY124187.1 | - | 10 - 11 | 1077 | 158 | |
AY124188.1 | - | 9 - 10 | 996 | 158 | |
AY124189.2 | - | 11 - 12 | 1151 | 158 | |
AY124190.1 | - | 10 - 11 | 1077 | 158 | |
AY124191.1 | - | 9 - 10 | 996 | 158 | |
AY124192.2 | - | 11 - 12 | 1151 | 158 | |
AY124193.1 | - | 10 - 11 | 1077 | 158 | |
AY124194.1 | - | 9 - 10 | 996 | 158 | |
BC108726.1 | - | 11 - 12 | 1095 | 158 | |
BC125217.1 | - | 8 - 9 | 792 | 158 | |
BC125218.1 | - | 8 - 9 | 792 | 158 | |
CR627441.1 | - | 11 - 12 | 1051 | 158 |