Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ROR2
- Entrez Gene ID
-
4920
- Gene Name
- receptor tyrosine kinase like orphan receptor 2
- Gene Aliases
-
BDB, BDB1, NTRKR2
- Chromosome Location
-
Chr.9: 91722596 - 91950206 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC033697.1 |
- |
9 - 10 |
2522 |
153 |
View Full Product Details