Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRTM4
Entrez Gene ID
80059
Gene Name
leucine rich repeat transmembrane neuronal 4
Gene Aliases
-
Chromosome Location
Chr.2: 76747723 - 77522433 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282928.1 NP_001269857.1 2 - 2 3109 139
NM_024993.4 NP_079269.4 3 - 3 3268 139
GenBank mRNA AK022630.1 - 1 - 1 2539 139
AK126961.1 - 3 - 3 3268 139
DB305042.1 - 1 - 1 139

View Full Product Details