Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FOXJ1
Entrez Gene ID
2302
Gene Name
forkhead box J1
Gene Aliases
FKHL13, HFH-4, HFH4
Chromosome Location
Chr.17: 76136333 - 76141299 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001454.3 NP_001445.2 1 - 2 184 84
GenBank mRNA BC046460.1 - 1 - 2 175 84
DB458983.1 - 1 - 2 184 84

View Full Product Details