The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000316.2 | NP_000307.1 | 1 - 2 | 95 | 62 |
XM_011533967.2 | XP_011532269.1 | 1 - 2 | 151 | 62 | |
XM_011533968.2 | XP_011532270.1 | 1 - 2 | 121 | 62 | |
XM_017006934.1 | XP_016862423.1 | 1 - 2 | 121 | 62 | |
GenBank mRNA | BC112221.1 | - | 1 - 2 | 95 | 62 |
BC112247.1 | - | 1 - 2 | 95 | 62 | |
U17418.1 | - | 1 - 2 | 110 | 62 | |
X68596.1 | - | 1 - 2 | 66 | 62 |