Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PDE1A
- Entrez Gene ID
-
5136
- Gene Name
- phosphodiesterase 1A
- Gene Aliases
-
CAM-PDE-1A, HCAM-1, HCAM1, HSPDE1A
- Chromosome Location
-
Chr.2: 182140035 - 182523375 on Build GRCh38
View Full Product Details