The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202502.1 | NP_001189431.1 | 7 - 8 | 565 | 66 |
NM_003634.3 | NP_003625.2 | 7 - 8 | 866 | 66 | |
GenBank mRNA | AJ001258.1 | - | 7 - 8 | 865 | 66 |
AK098367.1 | - | 7 - 8 | 620 | 66 | |
AK290953.1 | - | 7 - 8 | 619 | 66 | |
AK298816.1 | - | 7 - 8 | 567 | 66 | |
AK314407.1 | - | 7 - 8 | 637 | 66 | |
BC002371.2 | - | 7 - 8 | 616 | 66 | |
BC006473.1 | - | 7 - 8 | 616 | 66 | |
BC018869.2 | - | 6 - 7 | 508 | 66 |