The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198863.1 | NP_001185792.1 | 11 - 12 | 1148 | 62 |
NM_003915.5 | NP_003906.2 | 11 - 12 | 1375 | 62 | |
NM_152925.2 | NP_690902.1 | 11 - 12 | 1190 | 62 | |
NM_152926.2 | NP_690903.1 | 11 - 12 | 1148 | 62 | |
NM_152927.2 | NP_690904.1 | 12 - 13 | 1275 | 62 | |
NM_152928.2 | NP_690905.1 | 12 - 13 | 1243 | 62 | |
NR_037188.1 | 13 - 14 | 1367 | 62 | ||
GenBank mRNA | AB209827.1 | - | 11 - 12 | 1121 | 62 |
BC001142.1 | - | 13 - 14 | 1316 | 62 | |
BC021010.1 | - | 11 - 12 | 1097 | 62 | |
CR456790.1 | - | 10 - 11 | 993 | 62 | |
U83246.1 | - | 11 - 12 | 1149 | 62 |