Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLSPN
Entrez Gene ID
63967
Gene Name
claspin
Gene Aliases
-
Chromosome Location
Chr.1: 35720465 - 35769985 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190481.1 NP_001177410.1 12 - 13 2356 79
NM_022111.3 NP_071394.2 13 - 14 2548 79
XM_011541938.2 XP_011540240.1 13 - 14 2571 79
XM_011541939.2 XP_011540241.1 13 - 14 2565 79
XM_011541940.2 XP_011540242.1 13 - 14 2565 79
XM_011541941.2 XP_011540243.1 12 - 13 2403 79
GenBank mRNA AF297866.1 - 13 - 14 2548 79
BC113116.1 - 13 - 14 2541 79
BC115025.1 - 12 - 13 2349 79
BC115026.1 - 13 - 14 2541 79
BC137279.1 - 13 - 14 2527 79
BC140789.1 - 13 - 14 2527 79

View Full Product Details