The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190481.1 | NP_001177410.1 | 16 - 17 | 2928 | 83 |
NM_022111.3 | NP_071394.2 | 17 - 18 | 3120 | 83 | |
XM_011541938.2 | XP_011540240.1 | 17 - 18 | 3143 | 83 | |
XM_011541939.2 | XP_011540241.1 | 17 - 18 | 3137 | 83 | |
XM_011541940.2 | XP_011540242.1 | 17 - 18 | 3137 | 83 | |
XM_011541941.2 | XP_011540243.1 | 16 - 17 | 2975 | 83 | |
GenBank mRNA | AF297866.1 | - | 17 - 18 | 3120 | 83 |
BC113116.1 | - | 17 - 18 | 3113 | 83 | |
BC115025.1 | - | 16 - 17 | 2921 | 83 | |
BC115026.1 | - | 17 - 18 | 3113 | 83 | |
BC137279.1 | - | 17 - 18 | 3099 | 83 | |
BC140789.1 | - | 17 - 18 | 3099 | 83 |