The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166588.2 | NP_001160060.1 | 6 - 7 | 893 | 125 |
NM_001261418.1 | NP_001248347.1 | 6 - 7 | 893 | 125 | |
NM_020365.4 | NP_065098.1 | 6 - 7 | 893 | 125 | |
XM_017002745.1 | XP_016858234.1 | 6 - 7 | 781 | 125 | |
XM_017002746.1 | XP_016858235.1 | 7 - 8 | 860 | 125 | |
XM_017002747.1 | XP_016858236.1 | 7 - 8 | 854 | 125 | |
GenBank mRNA | AF257077.1 | - | 6 - 7 | 756 | 125 |
AK022399.1 | - | 6 - 7 | 755 | 125 | |
AK024006.1 | - | 6 - 7 | 781 | 125 | |
AK314668.1 | - | 6 - 7 | 766 | 125 | |
AL834288.1 | - | 6 - 7 | 758 | 125 | |
BC018728.1 | - | 6 - 7 | 751 | 125 | |
BC021017.1 | - | 6 - 7 | 744 | 125 | |
CR457285.1 | - | 2 - 3 | 147 | 125 |