The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198665.1 | NP_001185594.1 | 9 - 10 | 1055 | 82 |
NM_001301084.1 | NP_001288013.1 | 10 - 11 | 938 | 82 | |
NM_015313.2 | NP_056128.1 | 10 - 11 | 1112 | 82 | |
XM_006718805.3 | XP_006718868.1 | 10 - 11 | 1471 | 82 | |
XM_011542720.2 | XP_011541022.1 | 10 - 11 | 1053 | 82 | |
XM_017017421.1 | XP_016872910.1 | 10 - 11 | 1328 | 82 | |
GenBank mRNA | AB002380.2 | - | 5 - 6 | 474 | 82 |
AF180681.1 | - | 10 - 11 | 784 | 82 | |
AK294803.1 | - | 10 - 11 | 913 | 82 | |
AK304316.1 | - | 10 - 11 | 1336 | 82 | |
BC063117.1 | - | 9 - 10 | 1058 | 82 | |
BX647785.1 | - | 10 - 11 | 914 | 82 | |
BX648515.1 | - | 10 - 11 | 919 | 82 |