Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGEF12
Entrez Gene ID
23365
Gene Name
Rho guanine nucleotide exchange factor 12
Gene Aliases
LARG, PRO2792
Chromosome Location
Chr.11: 120336276 - 120489936 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198665.1 NP_001185594.1 30 - 31 3310 88
NM_001301084.1 NP_001288013.1 31 - 32 3193 88
NM_015313.2 NP_056128.1 31 - 32 3367 88
XM_006718805.3 XP_006718868.1 30 - 31 3651 88
XM_011542720.2 XP_011541022.1 31 - 32 3308 88
XM_017017420.1 XP_016872909.1 29 - 30 3823 88
XM_017017421.1 XP_016872910.1 31 - 32 3583 88
XM_017017422.1 XP_016872911.1 30 - 31 3075 88
GenBank mRNA AB002380.2 - 26 - 27 2729 88
AF180681.1 - 31 - 32 3039 88
AK294803.1 - 31 - 32 3168 88
AK304316.1 - 31 - 32 3591 88

View Full Product Details