The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198665.1 | NP_001185594.1 | 35 - 36 | 3809 | 134 |
NM_001301084.1 | NP_001288013.1 | 36 - 37 | 3692 | 134 | |
NM_015313.2 | NP_056128.1 | 36 - 37 | 3866 | 134 | |
XM_006718805.3 | XP_006718868.1 | 35 - 36 | 4150 | 134 | |
XM_011542720.2 | XP_011541022.1 | 36 - 37 | 3807 | 134 | |
XM_017017420.1 | XP_016872909.1 | 34 - 35 | 4322 | 134 | |
XM_017017421.1 | XP_016872910.1 | 36 - 37 | 4082 | 134 | |
XM_017017422.1 | XP_016872911.1 | 35 - 36 | 3574 | 134 | |
GenBank mRNA | AB002380.2 | - | 31 - 32 | 3228 | 134 |
AF180681.1 | - | 36 - 37 | 3538 | 134 | |
AK304316.1 | - | 36 - 37 | 4090 | 134 | |
BX647785.1 | - | 36 - 37 | 3668 | 134 |