The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243891.1 | NP_001230820.1 | 4 - 5 | 439 | 77 |
NM_001243895.1 | NP_001230824.1 | 4 - 5 | 514 | 77 | |
NM_003288.3 | NP_003279.2 | 4 - 5 | 514 | 77 | |
NM_199360.2 | NP_955392.1 | 4 - 5 | 514 | 77 | |
NM_199362.2 | NP_955394.1 | 4 - 5 | 514 | 77 | |
XM_011529034.2 | XP_011527336.1 | 4 - 5 | 514 | 77 | |
XM_011529035.2 | XP_011527337.1 | 4 - 5 | 514 | 77 | |
GenBank mRNA | AF004430.1 | - | 4 - 5 | 452 | 77 |
AK223031.1 | - | 4 - 5 | 426 | 77 | |
AK293349.1 | - | 4 - 5 | 336 | 77 | |
AK314501.1 | - | 4 - 5 | 472 | 77 | |
AU124850.1 | - | 4 - 5 | 443 | 77 | |
BC006804.1 | - | 4 - 5 | 399 | 77 | |
BI667351.1 | - | 3 - 4 | 266 | 77 | |
BM473873.1 | - | 4 - 5 | 471 | 77 | |
BT019631.1 | - | 4 - 5 | 376 | 77 | |
CR542036.1 | - | 4 - 5 | 376 | 77 | |
DA547106.1 | - | 4 - 5 | 415 | 77 | |
DC428780.1 | - | 4 - 5 | 369 | 77 |