The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016417.2 | NP_057501.2 | 1 - 2 | 402 | 77 |
GenBank mRNA | AB223038.1 | - | 1 - 2 | 341 | 77 |
BC023528.2 | - | 1 - 2 | 352 | 77 | |
BC047680.1 | - | 1 - 2 | 348 | 77 | |
BI259789.1 | - | 1 - 2 | 503 | 77 | |
BI834047.1 | - | 1 - 2 | 150 | 77 | |
BX248075.1 | - | 1 - 2 | 355 | 77 | |
DQ083331.1 | - | 1 - 2 | 298 | 77 |