Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PIGX
- Entrez Gene ID
-
54965
- Gene Name
- phosphatidylinositol glycan anchor biosynthesis class X
- Gene Aliases
-
PIG-X
- Chromosome Location
-
Chr.3: 196712374 - 196736007 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK294340.1 |
- |
3 - 4 |
663 |
101 |
View Full Product Details