Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFSWAP
Entrez Gene ID
6433
Gene Name
splicing factor SWAP homolog
Gene Aliases
SFRS8, SWAP
Chromosome Location
Chr.12: 131711087 - 131799738 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261411.1 NP_001248340.1 8 - 9 1462 123
NM_004592.3 NP_004583.2 8 - 9 1462 123
XM_011538653.1 XP_011536955.1 8 - 9 1459 123
XM_011538654.2 XP_011536956.1 8 - 9 1414 123
XM_011538655.2 XP_011536957.1 8 - 9 1459 123
XM_017019798.1 XP_016875287.1 8 - 9 1459 123
XM_017019799.1 XP_016875288.1 8 - 9 1901 123
GenBank mRNA AB208921.1 - 6 - 7 4653 123
BC008707.2 - 8 - 9 1459 123
BC015953.2 - 8 - 9 1459 123
BC024041.1 - 8 - 9 1460 123
BC113420.1 - 8 - 9 1365 123
BC136678.1 - 8 - 9 1412 123
BC144364.1 - 8 - 9 1412 123
BM454299.1 - - 248 123
U08377.1 - 8 - 9 1439 123

View Full Product Details