The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018051.4 | NP_060521.4 | 11 - 12 | 1620 | 60 |
XM_005249549.1 | XP_005249606.1 | 8 - 9 | 1212 | 60 | |
XM_006716041.2 | XP_006716104.1 | 11 - 12 | 1644 | 60 | |
XM_011516367.1 | XP_011514669.1 | 11 - 12 | 1486 | 60 | |
XM_011516368.2 | XP_011514670.1 | 11 - 12 | 1644 | 60 | |
XM_011516369.2 | XP_011514671.1 | 11 - 12 | 1475 | 60 | |
XM_017012381.1 | XP_016867870.1 | 11 - 12 | 1666 | 60 | |
XM_017012382.1 | XP_016867871.1 | 11 - 12 | 1635 | 60 | |
XM_017012383.1 | XP_016867872.1 | 11 - 12 | 1696 | 60 | |
XM_017012384.1 | XP_016867873.1 | 8 - 9 | 1203 | 60 | |
XM_017012385.1 | XP_016867874.1 | 10 - 11 | 1377 | 60 | |
XM_017012387.1 | XP_016867876.1 | 11 - 12 | 1496 | 60 | |
GenBank mRNA | AL137660.1 | - | 7 - 8 | 793 | 60 |
BC014491.1 | - | 11 - 12 | 1620 | 60 | |
CR627036.1 | - | 6 - 7 | 3262 | 60 |