The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018051.4 | NP_060521.4 | 18 - 19 | 2524 | 132 |
XM_005249549.1 | XP_005249606.1 | 15 - 16 | 2116 | 132 | |
XM_006716041.2 | XP_006716104.1 | 18 - 19 | 2548 | 132 | |
XM_011516367.1 | XP_011514669.1 | 18 - 19 | 2390 | 132 | |
XM_011516368.2 | XP_011514670.1 | 18 - 19 | 2548 | 132 | |
XM_011516369.2 | XP_011514671.1 | 18 - 19 | 2379 | 132 | |
XM_017012381.1 | XP_016867870.1 | 18 - 19 | 2570 | 132 | |
XM_017012382.1 | XP_016867871.1 | 18 - 19 | 2539 | 132 | |
XM_017012383.1 | XP_016867872.1 | 18 - 19 | 2600 | 132 | |
XM_017012384.1 | XP_016867873.1 | 15 - 16 | 2107 | 132 | |
XM_017012385.1 | XP_016867874.1 | 17 - 18 | 2281 | 132 | |
XM_017012386.1 | XP_016867875.1 | 15 - 16 | 2060 | 132 | |
XM_017012387.1 | XP_016867876.1 | 18 - 19 | 2400 | 132 | |
XM_017012388.1 | XP_016867877.1 | 8 - 9 | 1284 | 132 | |
GenBank mRNA | AK001162.1 | - | 2 - 3 | 239 | 132 |
AK027228.1 | - | 7 - 8 | 880 | 132 | |
AL137660.1 | - | 13 - 14 | 1524 | 132 | |
BC014491.1 | - | 18 - 19 | 2524 | 132 | |
BM718580.1 | - | 3 - 4 | 347 | 132 | |
CR627036.1 | - | 13 - 14 | 4166 | 132 |