The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018051.4 | NP_060521.4 | 19 - 20 | 2591 | 154 |
XM_005249549.1 | XP_005249606.1 | 16 - 17 | 2183 | 154 | |
XM_006716041.2 | XP_006716104.1 | 19 - 20 | 2615 | 154 | |
XM_011516367.1 | XP_011514669.1 | 19 - 20 | 2457 | 154 | |
XM_011516368.2 | XP_011514670.1 | 19 - 20 | 2615 | 154 | |
XM_011516369.2 | XP_011514671.1 | 19 - 20 | 2446 | 154 | |
XM_017012381.1 | XP_016867870.1 | 19 - 20 | 2637 | 154 | |
XM_017012382.1 | XP_016867871.1 | 19 - 20 | 2606 | 154 | |
XM_017012383.1 | XP_016867872.1 | 19 - 20 | 2667 | 154 | |
XM_017012384.1 | XP_016867873.1 | 16 - 17 | 2174 | 154 | |
XM_017012385.1 | XP_016867874.1 | 18 - 19 | 2348 | 154 | |
XM_017012386.1 | XP_016867875.1 | 16 - 17 | 2127 | 154 | |
XM_017012387.1 | XP_016867876.1 | 19 - 20 | 2467 | 154 | |
XM_017012388.1 | XP_016867877.1 | 9 - 10 | 1351 | 154 | |
GenBank mRNA | AK001162.1 | - | 3 - 4 | 306 | 154 |
AK027228.1 | - | 8 - 9 | 947 | 154 | |
AL137660.1 | - | 14 - 15 | 1591 | 154 | |
BC014491.1 | - | 19 - 20 | 2591 | 154 | |
BM718580.1 | - | 4 - 5 | 414 | 154 | |
CR627036.1 | - | 14 - 15 | 4233 | 154 |