The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018051.4 | NP_060521.4 | 6 - 7 | 1096 | 104 |
XM_006716041.2 | XP_006716104.1 | 6 - 7 | 1111 | 104 | |
XM_011516367.1 | XP_011514669.1 | 6 - 7 | 953 | 104 | |
XM_011516368.2 | XP_011514670.1 | 6 - 7 | 1111 | 104 | |
XM_011516369.2 | XP_011514671.1 | 6 - 7 | 942 | 104 | |
XM_017012381.1 | XP_016867870.1 | 6 - 7 | 1133 | 104 | |
XM_017012382.1 | XP_016867871.1 | 6 - 7 | 1111 | 104 | |
XM_017012383.1 | XP_016867872.1 | 6 - 7 | 1172 | 104 | |
XM_017012385.1 | XP_016867874.1 | 4 - 5 | 735 | 104 | |
XM_017012386.1 | XP_016867875.1 | 4 - 5 | 735 | 104 | |
XM_017012387.1 | XP_016867876.1 | 4 - 5 | 735 | 104 | |
GenBank mRNA | AL137660.1 | - | 2 - 3 | 269 | 104 |
BC014491.1 | - | 6 - 7 | 1096 | 104 | |
CR627036.1 | - | 1 - 2 | 49 | 104 |