Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DOK4
Entrez Gene ID
55715
Gene Name
docking protein 4
Gene Aliases
IRS-5, IRS5
Chromosome Location
Chr.16: 57471958 - 57486473 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018110.3 NP_060580.2 7 - 8 1086 59
XM_005256042.3 XP_005256099.1 6 - 7 1028 59
XM_005256043.2 XP_005256100.1 6 - 7 1028 59
GenBank mRNA AF466369.1 - 6 - 7 736 59
AK001350.1 - 7 - 8 1013 59
AK315426.1 - 6 - 7 996 59
BC001540.2 - 6 - 7 785 59
BC003541.1 - 7 - 8 1028 59

View Full Product Details