The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000074.2 | NP_000065.1 | 2 - 3 | 358 | 205 |
GenBank mRNA | AF529195.1 | - | 2 - 3 | 286 | 205 |
AK292637.1 | - | 2 - 3 | 350 | 205 | |
AY197739.1 | - | 2 - 3 | 286 | 142 | |
BC071754.1 | - | 2 - 3 | 358 | 205 | |
BC074950.2 | - | 2 - 3 | 321 | 205 | |
FN298241.1 | - | 2 - 3 | 297 | 205 | |
L07414.1 | - | 2 - 3 | 325 | 205 | |
X67878.1 | - | 2 - 3 | 331 | 205 | |
X68550.1 | - | 2 - 3 | 342 | 205 | |
X96710.1 | - | 2 - 3 | 307 | 205 | |
Z15017.1 | - | 2 - 3 | 307 | 205 |