Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC9A6
Entrez Gene ID
10479
Gene Name
solute carrier family 9 member A6
Gene Aliases
MRSA, NHE6
Chromosome Location
Chr.X: 135974597 - 136047269 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042537.1 NP_001036002.1 1 - 2 401 73
NM_001177651.1 NP_001171122.1 2 - 3 279 73
NM_006359.2 NP_006350.1 1 - 2 401 73
XM_006724726.3 XP_006724789.1 2 - 3 395 73
XM_011531243.2 XP_011529545.1 2 - 3 255 73
XM_017029223.1 XP_016884712.1 2 - 3 412 73
XM_017029224.1 XP_016884713.1 2 - 3 542 73
XM_017029225.1 XP_016884714.1 2 - 3 275 73
GenBank mRNA AB074255.1 - 1 - 2 325 73
AF030409.1 - 1 - 2 360 73
AK289775.1 - 1 - 2 325 73
AK296444.1 - 1 - 2 64 73
AK300475.1 - 2 - 3 270 73
AK314562.1 - 1 - 2 393 73
AK316255.1 - 2 - 3 270 73
BC035029.1 - 1 - 2 363 73
BC049169.1 - 1 - 2 325 73
D87743.1 - 1 - 2 316 73
DB483081.1 - 1 - 2 401 73
DC308430.1 - 2 - 3 280 73

View Full Product Details